Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haws™ Power Indicator Light Kit
SDP

Catalog No. 18888301 Shop All Haws Corporation Products
Change view
Click to view available options
For Use With (Equipment):
8317CTFP series Shower
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-888-301 8317CTFP series Shower
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-888-301 Supplier Haws™ Supplier No. 8317IDLTEXP
  • $1,261.00
    $943.65 / Each of 1

    Save $317.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Haws™ Power Indicator Light Kit is for use with Haws™ 8317CTFP series showers

Model 8317IDLTEXP is an indicator light kit for the 8317CTFP series showers. When lit, the light indicates power is being supplied to heat traced cable on model. Model is CSA electrical certified, TYPE 4. For Class 1, Div. 2, Group B, C & D. Tempering code T6; only when used with 8317CTFP.

Warranty and Services

1 year from ship date (Limited)

Specifications

Color Gray
For Use With (Equipment) 8317CTFP series Shower
Product Type Indicator Light
Weight (English) 2 lb.
Weight (Metric) 0.907185 kg
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.