Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haws™ Privacy Curtains
SDP

Catalog No. 17988387 Shop All Haws Corporation Products
Change view
Click to view available options
Product Type:
Curtain
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-988-387 Curtain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-988-387 Supplier Haws™ Supplier No. 9037
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Privacy gown

For horizontal emergency drench showers or drench shower and eyewash combinations

  • Wrap around design to enclose user
  • Corrosion resistant curtain rod
  • Partial water containment
  • 120.6cm (47.5 in.) curtain rod diameter
  • Made from Tyvek™
  • Color: yellow

Specifications

Color Yellow
For Use With (Equipment) Horizontal emergency drench shower or combination emergency shower/eyewash unit
Material Tyvek
Product Type Curtain
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.