Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haws™ Push Plate
SDP

Catalog No. 18888305 Shop All Haws Corporation Products
Change view
Click to view available options
For Use With (Equipment):
8300FP, Freeze-resistant MK Valves
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-888-305 8300FP, Freeze-resistant MK Valves
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-888-305 Supplier Haws™ Supplier No. SP218
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Haws™ Push Plate is for use with freeze-resistant MK valves.

Model SP218 is a single motion large zinc plated steel push plate for freeze-resistant valves. Valve stays open until manually closed.

Warranty and Services

1 year from ship date (Limited)

Specifications

Color Green
For Use With (Equipment) 8300FP, Freeze-resistant MK Valves
Material Zinc-plated Steel
Product Type Push Plate
Weight (English) 6 lb.
Weight (Metric) 2.72155 kg
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.