Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haws™ Replacement Dust Covers for Soft-Flo Spray Heads
SDP

Catalog No. 189992865 Shop All Haws Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Soft-Flo™ spray head
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-999-2865 Soft-Flo™ spray head
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-999-2865 Supplier Haws™ Supplier No. 9095
  • $75.20
    $52.65 / Pack of 1

    Save $22.55

    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fit ball type Soft-Flo spray heads.

Polyethylene Safety Green plastic covers with integral retainer strap protect eyewash spray heads from dust and debris.

  • Fit ball type Soft-Flo spray heads used on nonportable units 17-161-1A through -1D and 17-988-403; 17-988-386, 17-160, 17-160-1A through -1C and 19-033-727; 17-160-2
  • Dust covers automatically release when the water is turned on

Order Info

Sold one pair per pack.

Specifications

Color Green
For Use With (Equipment) Soft-Flo™ spray head
Includes Integral Retainer Strap
Material Plastic
Product Type Dust Cover
Weight (English) 0.05 lb.
Weight (Metric) 0.02 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.