Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haws™ Shower Test Kit
SDP

Catalog No. 18888318 Shop All Haws Corporation Products
Change view
Click to view available options
For Use With (Equipment):
8130, 8133H, Emergency Shower
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-888-318 8130, 8133H, Emergency Shower
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-888-318 Supplier Haws™ Supplier No. 9010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Haws™ Shower Test Kit is for use with emergency showers

Model 9010 shower test kit affords periodic shower testing with ease. The kit includes an upper corrosion resistant ring to encircle the shower head and tunnel shaped yellow Tyvek waterproof tube to guide the water column to model 9009 waste water container, or drain. Holding rod can be used as a handle for ease of testing by user. Testing record card included.

Warranty and Services

1 year from ship date (Limited)

Specifications

Color Yellow
For Use With (Equipment) 8130, 8133H, Emergency Shower
Height (English) 78 in.
Height (Metric) 198.12 cm
Material Plastic
Product Type Shower Test Kit
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.