Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haws™ Tank Cap
SDP

Catalog No. 18888327 Shop All Haws Corporation Products
Change view
Click to view available options
For Use With (Application):
For Use with Gravity Fed Eyewashes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
18-888-327 For Use with Gravity Fed Eyewashes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-888-327 Supplier Haws™ Supplier No. SP131
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Haws™ Tank Cap is a threaded black plastic filler cap for gravity fed eyewashes.

Model SP131 is a cap assembly with threaded cap and wide fill opening for easy filling, inspection and cleaning for the Haws gravity fed eyewashes.

Warranty and Services

1 year from ship date (Limited)

Specifications

Color Black
For Use With (Application) For Use with Gravity Fed Eyewashes
Material Plastic
Product Type Tank Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.