Learn More
Haws™ Thermal Protection Jackets
SDP
Catalog No. 19811867 Shop All Haws Corporation Products
Change view
Click to view available options
Product Type:
Thermal Protection Jacket
Catalog No. Product Type
19-811-867 Thermal Protection Jacket
1 options
Catalog No. 19-811-867 Supplier Haws™ Supplier No. 9125
  • $1,810.00
    $1,355.65 / Each of 1
    Save $454.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Model 9125 is a white thermal protection jacket for 7500 series gravity eyewashes.
Model 9125 is a thermal protection jacket. Recommended for areas where extreme heat or sunlight might cause water to overheat
Specifications
Product Type Thermal Protection Jacket
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov