Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haws™ Thermostat
SDP

Catalog No. 18888335 Shop All Haws Corporation Products
Change view
Click to view available options
For Use With (Equipment):
8317CTFP and 8317CTFP.220V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-888-335 8317CTFP and 8317CTFP.220V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-888-335 Supplier Haws™ Supplier No. SP155
Add to Cart
Add to Cart

Haws™ Thermostat is for the heat traced cable outdoor Models 8317CTFP and 8317CTFP.220V

Model SP155 is a thermostat for the heat traced cable outdoor Models 8317CTFP and 8317CTFP.220V. It has hermetically sealed contacts. Opens (rising) at 62°F (17°C) and closes (falling) at 52°F (11°C).

Warranty and Services

1 year from ship date (Limited)

Specifications

Certifications/Compliance ANSI Z358.1
Color Black, White
For Use With (Application) Model SP155 is a thermostat for the heat traced cable outdoor Models 8317CTFP and 8317CTFP.220V
For Use With (Equipment) 8317CTFP and 8317CTFP.220V
Product Type Thermostat
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.