Learn More
Haws™ Waste Container
SDP
Catalog No. 18888358 Shop All Haws Corporation Products
Change view
Click to view available options
Capacity (English):
20 gal.
Catalog No. Capacity (English)
18-888-358 20 gal.
1 options
Catalog No. 18-888-358 Supplier Haws™ Supplier No. 9009
  • $376.58
    $271.65 / Each of 1
    Save $104.93
    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Haws™ Waste Container is used to collect water during weekly testing of emergency equipment.
Model 9009 waste water container adapts to model 9008 to be used with models 7500 or 9010. Twenty gallon (75.7 L) yellow container constructed of tough polymer resin.
Warranty and Services
1 year from ship date (Limited)
Specifications
Capacity (Metric) 75.7 L
Certifications/Compliance ANSI Z358.1
Capacity (English) 20 gal.
Color Yellow
Material Polymer Resin
Product Type Waste Container