Learn More
Aviva Systems Biology Corp HAX1 Antibody - middle region (ARP52142_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52142_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HAX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HAX1. Alternate names: HCLSBP1, HS1BP1, SCN3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-648