Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hayem Diluting Fluid, for Erythrocyte (Red Cell) Count, Ricca Chemical
SDP

Catalog No. 35004 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3500-4 120 mL Amber Glass Bottle
3500-16 500 mL Amber Glass Bottle
3500-32 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 3500-4 Supplier Ricca Chemical Company Supplier No. 35004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Erythrocyte (Red Cell) Count

Specifications

Physical Form Liquid
Chemical Name or Material Hayem Diluting Fluid
CAS 7732-18-5, 7757-82-6, 7647-14-5, 7487-94-7
CAS Min % 95.0, 2.37, 0.47, 0.24
CAS Max % 99.0, 2.47, 0.49, 0.25
Packaging Amber Glass Bottle
Color Colorless
Quantity 120 mL
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.