Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ HayeSep™ Porous Polymer Adsorbent, Matrix HayeSep™ P, 60-80 mesh

Catalog No. 111004195 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
75 cc
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4195 75 cc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4195 Supplier MilliporeSigma™ Supelco™ Supplier No. 10297
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HayeSep™ P column was used for the gas chromatographic analysis of the vapour phase in an experiment conducted to know the solubility of weak electrolytes such as ammonia, carbon dioxide, sulphur dioxide and hydrogen cyanide for process design in various applications.

HayeSep™ porous polymers, considered second generation materials, are consistent batch-to-batch, with minimal shrinkage and monomer bleed.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 60/80 Mesh
Packaging Bottle
Quantity 75 cc
Surface Area (Metric) ˜165 m2/g
Product Line HayeSep
For Use With (Application) GC
Density ˜0.42 g/mL (free fall density)

HayeSep is a registered trademark of Hayes Separation Inc.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.