Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Haz Loc Cord Reel 50ft 14 AWG

Catalog No. 502769803
Encompass_Preferred

Haz Loc Cord Reel 50ft 14 AWG Bare 600V

Catalog No. 50-276-9803 Supplier Grainger Supplier No. 13V224
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Haz Loc Cord Reel 50ft 14 AWG Bare 600V

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.