Learn More
Andwin Scientific Hazardous Warning Labels, 4x7
SDP
Catalog No. NC9522200
Encompass_Preferred
Hazardous Warning Labels, Hand write the chemical name, fill in the warning diamond and circle the appropriate personal protection symbols. A self-laminating flap then seals the writing and protects the label from solvents, acids, etc.
Catalog No. NC9522200 Supplier Andwin Scientific Supplier No. 36433437
Hazardous Warning Labels, Hand write the chemical name, fill in the warning diamond and circle the appropriate personal protection symbols. A self-laminating flap then seals the writing and protects the label from solvents, acids, etc.