Learn More
Labelmaster Inc Hazardous Waste Label, Paper Stock
SDP
Catalog No. NC9794832
Encompass_Preferred
Hazardous Waste Accumulation Labels provide a convenient way for a generator to mark accumulation containers in accordance with EPA regulations in 40 CFR part 262.34. Paper Hazardous Waste Labels offer an economical alternative to vinyl.
Catalog No. NC9794832 Supplier Labelmaster Inc Supplier No. HWA
Hazardous Waste Accumulation Labels provide a convenient way for a generator to mark accumulation containers in accordance with EPA regulations in 40 CFR part 262.34. Paper Hazardous Waste Labels offer an economical alternative to vinyl.