Learn More
Labelmaster Inc Dry Ice Label, Paper, Roll of 500
SDP
Catalog No. NC0763856
Encompass_Preferred
Labelmaster’s paper, 6" x 6" Worded Dry Ice Labels have UN1845 Dry Ice preprinted on them. There are blanks to fill in your dry ice quantity, the shipper's name and the consignee. These Worded Dry Ice Labels meet the design and durability standards of 49 CFR, ICAO and other international air regulations. The Hazard Class 9 labels can help you stay in compliance with standards set forth by the DOT. Hazmat labels are printed with light-fast inks for high durability, and adhesives are formulated for use in various climates and environments.
Catalog No. NC0763856 Supplier Labelmaster Inc Supplier No. HML11DICE
Labelmaster’s paper, 6" x 6" Worded Dry Ice Labels have UN1845 Dry Ice preprinted on them. There are blanks to fill in your dry ice quantity, the shipper's name and the consignee. These Worded Dry Ice Labels meet the design and durability standards of 49 CFR, ICAO and other international air regulations. The Hazard Class 9 labels can help you stay in compliance with standards set forth by the DOT. Hazmat labels are printed with light-fast inks for high durability, and adhesives are formulated for use in various climates and environments.