Learn More
MSC TPI Industrial Drum Fan, 42 in., 3/4 hp, 120/240 V, 1 ea.
Catalog No. 5000507298
Encompass
This 42 in. industrial belt drive portable blower is designed for use in hazardous locations, featuring an explosion-proof motor and construction compliant with Class I, Group D and Class II, Groups E, F, and G ratings. It is engineered for effective air circulation in demanding industrial environments, schools, and large warehouse spaces.
  • Equipped with an explosion-proof motor and external junction box for hazardous location safety
  • Meets OSHA standards for workplace safety
  • Features 4-paddle aluminum blades
  • Includes spiral wire front and rear guards for protection
  • Operates at 1-speed with a 3/4 hp motor
  • Hardwired electrical connection required
  • Constructed with a black housing and integrated 10 in. wheels for mobility
Catalog No. 50-005-07298 Supplier MSC Supplier No. 77991537
This 42 in. industrial belt drive portable blower is designed for use in hazardous locations, featuring an explosion-proof motor and construction compliant with Class I, Group D and Class II, Groups E, F, and G ratings. It is engineered for effective air circulation in demanding industrial environments, schools, and large warehouse spaces.
  • Equipped with an explosion-proof motor and external junction box for hazardous location safety
  • Meets OSHA standards for workplace safety
  • Features 4-paddle aluminum blades
  • Includes spiral wire front and rear guards for protection
  • Operates at 1-speed with a 3/4 hp motor
  • Hardwired electrical connection required
  • Constructed with a black housing and integrated 10 in. wheels for mobility