Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC TPI Suspension Blower, Hazardous Location, 48 in., 1 HP, 1 ea

Catalog No. 5000507440
Encompass

TPI Suspension Blower, Hazardous Location, 48 in., 1 HP, 1 ea

Catalog No. 50-005-07440 Supplier MSC Supplier No. 77991602
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TPI Suspension Blower, Hazardous Location, 48 in., 1 HP, 1 ea

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.