Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Hazard Marking Anti-Skid Tape Roll - Anti-Skid Laminated Vinyl, CAUTION DO NOT ENTER, Black on Yellow

Catalog No. 19103822 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black on Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-103-822 Black on Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-103-822 Supplier Brady™ Supplier No. 92415
  • $183.00
    $115.65 / Each of 1

    Save $67.35

    37% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

B-819 Anti-Skid Vinyl features a velvet embossed rigid overlaminate

  • Excellent chemical, oil and water resistance
  • Anti-skid hazard marking tape identifies potential hazards while also helping to prevent slips, trips and falls in wet or high traffic areas

Specifications

Color Black on Yellow
Width (Metric) 140.97 mm
Width (English) 3 in.
Certifications/Compliance ANSI A1264.2-2006, NSFI Certified for High Traction, OSHA 1910.144, OSHA 1910.22
Thickness 0.012 in.
Displayed Text CAUTION DO NOT ENTER
Length (English) 18 yd.
Packaging Quantity 54 ft./Roll
Material Vinyl
Type Anti-Skid Tape
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.