Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Hazard Marking Anti-Skid Tape Roll - Anti-Skid Laminated Vinyl, CAUTION WATCH YOUR STEP, Black on Yellow
Shop All Brady Corporation ProductsDescription
- Excellent chemical, oil and water resistance
- Anti-skid hazard marking tape identifies potential hazards while also helping to prevent slips, trips and falls in wet or high traffic areas
Specifications
Specifications
| Color | Black on Yellow |
| Certifications/Compliance | ANSI A1264.2-2006, NSFI Certified for High Traction, OSHA 1910.144, OSHA 1910.22 |
| Width (English) | 3 in. |
| Type | Anti-Skid Tape |
| Displayed Text | CAUTION WATCH YOUR STEP |
| Length (English) | 18 yd. |
| Packaging Quantity | 54 ft./Roll |
| Thickness | 0.012 in. |
| Width (Metric) | 140.97 mm |
| Material | Vinyl |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?