Learn More
MSC TPI Industrial Circulator, 30 in., 1/4 hp, 3-Phase, 1 ea.
Catalog No. 5000507401
Encompass
This 30 in. hazardous location circulator fan head is designed for demanding industrial environments. It features a robust aluminum blade system and is engineered to meet stringent safety requirements in hazardous areas.
  • Hazardous location rated: Class I Group D and Class II Group F and G
  • Durable construction featuring steel hub and spider with aluminum paddle blades
  • Coated steel guards for added protection
  • Direct drive motor with ball bearing design
  • Totally enclosed and permanently lubricated motor
  • Meets Federal OSHA standards
  • Designed for hard wiring (no cord-junction box provided)
Catalog No. 50-005-07401 Supplier MSC Supplier No. 77993681
This 30 in. hazardous location circulator fan head is designed for demanding industrial environments. It features a robust aluminum blade system and is engineered to meet stringent safety requirements in hazardous areas.
  • Hazardous location rated: Class I Group D and Class II Group F and G
  • Durable construction featuring steel hub and spider with aluminum paddle blades
  • Coated steel guards for added protection
  • Direct drive motor with ball bearing design
  • Totally enclosed and permanently lubricated motor
  • Meets Federal OSHA standards
  • Designed for hard wiring (no cord-junction box provided)