Learn More
GEMPLERS INC HAZARDOUS LIQUID SPILL KIT
SDP
Catalog No. NC1896168
Encompass
This convenient Hazardous Liquid Spill Kit is ideal for a quick response to emergency leaks and spills. The plastic pail has a snap-on lid that keeps contents clean and dry, and also doubles as a disposal container once spills are cleaned up. Liquid Spill Kit contains: (2) Absorbent socks (10) Universal perforated pads (1) Disposal bag and tie.
Catalog No. NC1896168 Supplier GEMPLERS INC Supplier No. 145760
This convenient Hazardous Liquid Spill Kit is ideal for a quick response to emergency leaks and spills. The plastic pail has a snap-on lid that keeps contents clean and dry, and also doubles as a disposal container once spills are cleaned up. Liquid Spill Kit contains: (2) Absorbent socks (10) Universal perforated pads (1) Disposal bag and tie.