Learn More
MSC TPI Hazardous Location Snap Action Thermostat, Cast Aluminum, 50°F to 90°F, 22 A, 1 ea.
Catalog No. 5000507445
Encompass
This explosion-resistant, hazardous location snap action thermostat is designed for industrial and commercial heating and cooling applications. It features a robust 1/2-inch thick cast aluminum housing to maintain safety in challenging environments. The unit utilizes a bi-metal sensor for accurate room temperature regulation between 50°F to 90°F (10°C to 32°C).
  • Explosion-proof design for hazardous locations
  • Double pole double throw (DPDT) snap action switch
  • Rated for 22 A at 120-277 V AC resistive loads
  • Includes 3/4-inch conduit taps at the top and bottom
  • Compatible with single-phase hazardous location wall convectors
  • Includes 3/4-inch to 1/2-inch conduit adapter and plug
  • Certified for Class I, Group C and D and Class II, Group E, F, and G environments
Catalog No. 50-005-07445 Supplier MSC Supplier No. 76942572
This explosion-resistant, hazardous location snap action thermostat is designed for industrial and commercial heating and cooling applications. It features a robust 1/2-inch thick cast aluminum housing to maintain safety in challenging environments. The unit utilizes a bi-metal sensor for accurate room temperature regulation between 50°F to 90°F (10°C to 32°C).
  • Explosion-proof design for hazardous locations
  • Double pole double throw (DPDT) snap action switch
  • Rated for 22 A at 120-277 V AC resistive loads
  • Includes 3/4-inch conduit taps at the top and bottom
  • Compatible with single-phase hazardous location wall convectors
  • Includes 3/4-inch to 1/2-inch conduit adapter and plug
  • Certified for Class I, Group C and D and Class II, Group E, F, and G environments