Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Hazardous Material Label: BENZALDEHYDE

Catalog No. 19107628 Shop All Brady Corporation Products
Change view
Click to view available options
Display Text:
BENZALDEHYDE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text
19-107-628 BENZALDEHYDE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-107-628 Supplier Brady™ Supplier No. 93493

Provide workers with all of the chemical information needed for hazardous materials they may encounter. Brady™ Hazardous Material Labels are made of vinyl and display text in black on yellow.

  • Provides workers with chemical information of hazardous materials
  • Made from B-928 vinyl material
  • Label Size: W x H: 4.5 x 3.75 in.

Specifications

Adhesive Type Acrylic, Permanent, Cold-temperature
Color Black, Yellow
Material Vinyl (B-928)
Product Type Hazardous Materials Label
Display Text BENZALDEHYDE
Quantity 25/Pk.
Shape Rectangle
Size (English) 4.5 x 3.75 in. (W x H)
Thickness (English) 0.0049 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.