Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Hazardous Material Label, Legend: CONTAINS ASBESTOS…...CANCER AND LUNG DISEASE HAZARD

Catalog No. 19104935 Shop All Brady Corporation Products
Change view
Click to view available options
Size (English):
4 x 4 in.
6 x 6 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (English)
19-104-935 6 x 6 in.
19-104-920 4 x 4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-104-935 Supplier Brady™ Supplier No. 121438
  • $138.00
    $106.65 / Pack of 100

    Save $31.35

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to identify hazardous material storage. Brady™ Hazardous Material Labels send a clear message to workers when asbestos is present.

  • Abrasion and chemical resistant
  • Does not include Write-On-Area

Specifications

Adhesive Type Acrylic, Permanent
Color Black, Red, White
Finish Gloss
Material Vinyl Film (B-7569)
Product Type Hazardous Materials Label
Display Text DANGER: CONTAINS ASBESTOS FIBERS AVOID CREATING DUST CANCER AND LUNG DISEASE HAZARD
Shape Square
Size (English) 6 x 6 in.
Size (Metric) 15.24 x 15.24 cm
Thickness (English) 0.004 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.