Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Hazardous Waste Federal Law Prohibits Improper Disposal...etc Sign

Catalog No. 19000652 Shop All Brady Corporation Products
Change view
Click to view available options
Quantity:
100/Pk.
50/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-000-652 50/Pk.
19-110-971 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-000-652 Supplier Brady™ Supplier No. 121155
Add to Cart
Add to Cart

Identify containers, cylinders and drums holding hazardous waste. Brady™ Biohazard Sign help notify and warn employees about potentially dangerous materials in the facility, as required by industry regulations.

Comply with all current Federal regulations regarding identification of hazardous waste material for trasportation off-site

Specifications

Material Vinyl
Quantity 50/Pk.
Height (English) 6 in.
Height (Metric) 15.2 cm
Width (English) 6 in.
Width (Metric) 15.2 cm
Displayed Text HAZARDOUS WASTE FEDERAL LAW PROHIBITS IMPROPER DISPOSAL...etc
Product Type Hazardous Waste Label
Color Red on Yellow

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.