Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Hazardous Waste Label: NON-REGULATED WASTE

Catalog No. 19110977 Shop All Brady Corporation Products
Change view
Click to view available options
Quantity:
100/Pk.
50/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-110-977 50/Pk.
19-110-973 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-110-977 Supplier Brady™ Supplier No. 121158
  • $90.00
    $54.65 / Pack of 50

    Save $35.35

    39% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Identify hazardous waste materials for transportation off-site. Brady™ Hazardous Waste Labels are made of vinyl and comply with all current federal regulations regarding identification of hazardous waste material transportation.

  • To identify containers, cylinders and drums holding hazardous waste
  • Made of B-7569 vinyl
  • Available in size (W x H): 15.2 x 15.2cm (6 x 6 in.)

Specifications

Adhesive Type Acrylic, Permanent
Color White, Blue
Material Vinyl (B-7569)
Product Type Hazardous Waste Label
Display Text NON-REGULATED WASTE
Quantity 50/Pk.
Shape Square
Size (English) 6 x 6 in.
Thickness (English) 0.004 in.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.