Learn More
Description
- Color-coded yellow for safety
- Warns other in the area of a potentially hazardous spill or leak
- Heavyweight product provides additional absorbency and strength
- Perforated for flexibility and efficiency
- Great wicking and performance for all of your leaks, drips and spills.
- Dimple bond pattern increases strength and durability
Specifications
Specifications
| Color | Yellow |
| Capacity (English) | 18 gal. |
| Capacity (Metric) | 68 L |
| Material | Meltblown Polypropylene |
| Disposable | Single-use |
| Grade | Heavyweight |
| Packaging Quantity | 100/Bag |
| Dimensions (L x W) | 15 x 18 in. (38.1 x 45.7 cm) |
| Weight (English) | 10 lb. |
| Weight (Metric) | 4.5 kg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.