Learn More
Grainger Hazardous Waste Bags: 10 gal Capacity, 17 in Wd, 30 in Ht, Polyethylene, 24 PK
Catalog No. 5001028
Encompass_Preferred
Bags for hazardous waste are used to collect items that are contaminated with materials that can harm workers or damage the environment, including heavy metals, toxic chemicals, and infectious medical waste. They are used in industrial processing settings, chemotherapy clinics, and other settings where toxic or hazardous waste is produced.
This item is not returnable. View return policy
Catalog No. 50-010-28 Supplier Grainger Supplier No. 3WNA4
Bags for hazardous waste are used to collect items that are contaminated with materials that can harm workers or damage the environment, including heavy metals, toxic chemicals, and infectious medical waste. They are used in industrial processing settings, chemotherapy clinics, and other settings where toxic or hazardous waste is produced.