Learn More
Crystal Chem Inc Control for the Mouse HbA1c Assay Kit
SDP
Catalog No. 501947689
Encompass_Preferred
The Control for the Mouse HbA1c Assay can be used to validate the calibration curve obtained when running the Mouse HbA1c Assay Assay. Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.
Catalog No. 50-194-7689 Supplier Crystal Chem Inc Supplier No. 80313
The Control for the Mouse HbA1c Assay can be used to validate the calibration curve obtained when running the Mouse HbA1c Assay Assay. Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.