Learn More
Medchemexpress LLC HY-P7017 10ug Medchemexpress, HB-EGF, Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP701710UG
Encompass_Preferred
Medchemexpress, HY-P7017 10ug HB-EGF, Human CAS: Recombinant Human Heparin-binding EGF-like Growth Factor is a member of the epidermal growth factor (EGF) family of growth factors that stimulate growth and differentiation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4270