Learn More
Genscript Corporation Antibody Protein Proteins Proteins 1 MG 1/EA
Supplier:  Genscript Corporation Z032911
Encompass
Proheparin-binding EGF-like growth factor (HB-EGF) also known as DTR DTS and HEGFL is a member of the EGF family of mitogens. It is expressed in macrophages monocytes endothelial cells and muscle cells. HB-EGF signals through the EGF receptor to stimulate the proliferation of smooth muscle cells epithelial cells and keratinocytes. Compared to EGF HB-EGF binds to the EGF receptor with a higher affinity and has been shown to bemore mitogenic likely due to its ability to bind to heparin and heparin sulfate proteoglycans. HB-EGF has also been reported to act as a diphtheria toxin receptor mediating endocytosis of the bound toxin. Heparin-binding EGF-like growth factor has been shown to interact with NRD1 Zinc finger and BTB domain-containing protein 16 and BAG1.
Catalog No. 50-252-9690