Learn More
Medchemexpress LLC HB-EGF Human CHO 50ug
SDP
Catalog No. 5000179127
Encompass_Preferred
HB-EGF Protein Human (CHO) is shown to play a role in wound healing cardiac hypertrophy and heart development and function
Catalog No. 50-001-79127 Supplier Medchemexpress LLC Supplier No. HYP740050UG
HB-EGF Protein Human (CHO) is shown to play a role in wound healing cardiac hypertrophy and heart development and function