Learn More
MSC Fowler 54-100-312 0 to 300mm Range, 0.01mm Resolution, Electronic Caliper 3-1/2" Jaws, 0.04mm Accuracy, Serial Output
Catalog No. NC1816987
Encompass
Fowler 54-100-312 0 to 300mm Range, 0.01mm Resolution, Electronic Caliper 3-1/2" Jaws, 0.04mm Accuracy, Serial Output
Catalog No. NC1816987 Supplier MSC Supplier No. 65289852
Fowler 54-100-312 0 to 300mm Range, 0.01mm Resolution, Electronic Caliper 3-1/2" Jaws, 0.04mm Accuracy, Serial Output