Learn More
INTACT GENOMICS INC HB101 Chemically Competent Cells 6x100µl
Supplier:  INTACT GENOMICS INC 107106
Encompass
Intact Genomics (ig) HB101 chemically competent E. coli cells are suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning. E. coli HB101 is a K12 x B hybrid strain, containing the recA13 mutation that minimizes recombination and helps insert stability. In addition, it carries the hsdS20(rB-mB-) restriction minus genotype which prevents cleavage of cloned DNA by endogenous restriction enzymes. HB101 strain does not support Alpha-complementation for blue/white screening.Reagents Needed for One Reaction:ig HB101 chemically competent cells: 50 µlDNA (or pUC19 Control, 10 pg/µl): 1 µlRecovery medium: 1 ml
Catalog No. 50-000-82795