Learn More
Gold Biotechnology Inc HB101 Competent E coli Cells
SDP
Catalog No. 5000562229
Encompass_Preferred
GoldBio s HB101 chemically competent E coli cells allow you to obtain high transformation efficiency in applications such cloning and sub-cloning Our E coli HB101 proceeds from the K12 x B hybrid strain HB101 strain also contains the recA13 mutation useful in the insert stability and minimization of recombination E coli HB101 also has the hsdS20(rB-mB-) genotype that prevents the cleavage of cloned DNA by endogenous restriction enzymes
Catalog No. 50-005-62229 Supplier Gold Biotechnology Inc Supplier No. CC15010X50
GoldBio s HB101 chemically competent E coli cells allow you to obtain high transformation efficiency in applications such cloning and sub-cloning Our E coli HB101 proceeds from the K12 x B hybrid strain HB101 strain also contains the recA13 mutation useful in the insert stability and minimization of recombination E coli HB101 also has the hsdS20(rB-mB-) genotype that prevents the cleavage of cloned DNA by endogenous restriction enzymes