Learn More
Medchemexpress LLC HY-133520 5mg , HBC620 CAS:2530162-07-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1335205MG
Encompass_Preferred
HY-133520 5mg HBC620 CAS: 2530162-07-1 HBC620 is a HBC analog. HBC is nonfluorescent in solution, but emits strong fluorescence upon forming tight complex with Pepper RNA aptamer. HBC-Pepper complex can be used to visualize RNA dynamics in live cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9699