Learn More
Selleck Chemical LLC HBC620-E0623-25MG
SDP
Catalog No. 503134432
Encompass_Preferred
HBC620 is an analogue of HBC which is a GFP fluorophore-like synthetic dye that is nonfluorescent in solution but strongly fluoresces upon forming tight complex with Pepper RNA aptamer HBC-Pepper complex can be used to visualize RNA dynamics in live cells
Catalog No. 50-313-4432 Supplier Selleck Chemical LLC Supplier No. E062325MG
HBC620 is an analogue of HBC which is a GFP fluorophore-like synthetic dye that is nonfluorescent in solution but strongly fluoresces upon forming tight complex with Pepper RNA aptamer HBC-Pepper complex can be used to visualize RNA dynamics in live cells