Learn More
Enzo Life Sciences HBDDE (5mg). CAS: 154675-18-0
SDP
Supplier:  Enzo Life Sciences BMLEI2730005
Encompass_Preferred
Inhibits PKCα (IC50=43 µM) and PKCγ (IC50=50 µM) without inhibiting the δ, βI and βII isotypes. Purity: ≥95% (TLC). Solubility: Soluble in DMSO (25 mg/ml), ethanol (15 mg/ml warm) or water (10 mg/ml warm). Long Term Storage: -20°C.
Catalog No. 50-201-0654