Learn More
Medchemexpress LLC Hbed-cc-tris(tert-butyl ester) | 2097123-80-1 | 98.4% | 700.86 | C38H56N2O10 | 10 MG
SDP
Catalog No. 5000286344
Encompass_Preferred
HBED-CC-tris(tert-butyl ester) is a bifunctional chelating agent used in radiolabeling research and development of tumor-targeted diagnostic compounds. It is commonly conjugated to targeting vectors such as PSMA and labeled with gallium isotopes for in vitro studies supporting prostate cancer diagnostics. The material is supplied as a solid, white to off-white powder and is soluble in DMSO.
  • High purity (98.4%).
  • Bifunctional chelator compatible with common radiometal labeling workflows.
  • Soluble in DMSO for convenient formulation.
  • Stable as a powder under frozen storage for long-term use.
  • Suitable for conjugation to targeting ligands for diagnostic research.
Catalog No. 50-002-86344 Supplier Medchemexpress LLC Supplier No. HY15355110MG
HBED-CC-tris(tert-butyl ester) is a bifunctional chelating agent used in radiolabeling research and development of tumor-targeted diagnostic compounds. It is commonly conjugated to targeting vectors such as PSMA and labeled with gallium isotopes for in vitro studies supporting prostate cancer diagnostics. The material is supplied as a solid, white to off-white powder and is soluble in DMSO.
  • High purity (98.4%).
  • Bifunctional chelator compatible with common radiometal labeling workflows.
  • Soluble in DMSO for convenient formulation.
  • Stable as a powder under frozen storage for long-term use.
  • Suitable for conjugation to targeting ligands for diagnostic research.