Learn More
Medchemexpress LLC HBED-CC-tris(tert-butyl ester) | 2097123-80-1 | C38H56N2O10 | 10 MM * 1 ML
SDP
Catalog No. 5000373327
Encompass_Preferred
HBED-CC-tris(tert-butyl ester) is a derivative of HBED-CC that functions as a bifunctional chelator. It is utilized in research for radiolabeling and compounds for tumor-assisted diagnosis. This compound can also be used in research on the auxiliary diagnosis of prostate cancer when conjugated with PSMA and labeled with gallium isotopes.
  • Functions as a bifunctional chelator
  • Utilized in research for radiolabeling
  • Used in compounds for tumor-assisted diagnosis
  • Can be employed in research related to the auxiliary diagnosis of prostate cancer when conjugated with PSMA and labeled with gallium isotopes
Catalog No. 50-003-73327 Supplier Medchemexpress LLC Supplier No. HY15355110MM/1ML
HBED-CC-tris(tert-butyl ester) is a derivative of HBED-CC that functions as a bifunctional chelator. It is utilized in research for radiolabeling and compounds for tumor-assisted diagnosis. This compound can also be used in research on the auxiliary diagnosis of prostate cancer when conjugated with PSMA and labeled with gallium isotopes.
  • Functions as a bifunctional chelator
  • Utilized in research for radiolabeling
  • Used in compounds for tumor-assisted diagnosis
  • Can be employed in research related to the auxiliary diagnosis of prostate cancer when conjugated with PSMA and labeled with gallium isotopes