Learn More
Medchemexpress LLC HBED-CC-tris(tert-butyl ester) | 2097123-80-1 | C38H56N2O10 | 50 MG
SDP
Catalog No. 5000372715
Encompass_Preferred
HBED-CC-tris(tert-butyl ester) is a derivative of HBED-CC and a bifunctional chelator. It can be utilized in the research of radiolabeling and compounds for tumor-assisted diagnosis. Specifically, when conjugated with PSMA and labeled with gallium isotopes, it can be employed in research for the auxiliary diagnosis of prostate cancer.
  • Derivative of HBED-CC
  • Bifunctional chelator
  • Used in radiolabeling research
  • Used in compounds for tumor-assisted diagnosis
  • Employed in research for auxiliary diagnosis of prostate cancer when conjugated with PSMA and labeled with gallium isotopes
Catalog No. 50-003-72715 Supplier Medchemexpress LLC Supplier No. HY153551A50MG
HBED-CC-tris(tert-butyl ester) is a derivative of HBED-CC and a bifunctional chelator. It can be utilized in the research of radiolabeling and compounds for tumor-assisted diagnosis. Specifically, when conjugated with PSMA and labeled with gallium isotopes, it can be employed in research for the auxiliary diagnosis of prostate cancer.
  • Derivative of HBED-CC
  • Bifunctional chelator
  • Used in radiolabeling research
  • Used in compounds for tumor-assisted diagnosis
  • Employed in research for auxiliary diagnosis of prostate cancer when conjugated with PSMA and labeled with gallium isotopes