Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HBED-CC-tris(tert-butyl ester) TFA | C40H57F3N2O12 | 1 ML
SDP

Catalog No. 5000373841
Encompass_Preferred

HBED-CC-tris(tert-butyl ester) TFA is a derivative of HBED-CC and a bifunctional chelator. It is utilized in radiolabeling research and for compounds in tumor-assisted diagnosis. When conjugated with PSMA and labeled with gallium isotopes, it can be applied in research for the auxiliary diagnosis of prostate cancer.

  • Derivative of HBED-CC and a bifunctional chelator
  • Used in radiolabeling research
  • Supports research for tumor-assisted diagnosis
  • Purity of 99.50%
  • White to off-white solid appearance
  • Molecular weight of 814.88
  • Soluble in DMSO at concentrations of ≥ 100 mg/mL

Catalog No. 50-003-73841 Supplier Medchemexpress LLC Supplier No. HY153551A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HBED-CC-tris(tert-butyl ester) TFA is a derivative of HBED-CC and a bifunctional chelator. It is utilized in radiolabeling research and for compounds in tumor-assisted diagnosis. When conjugated with PSMA and labeled with gallium isotopes, it can be applied in research for the auxiliary diagnosis of prostate cancer.

  • Derivative of HBED-CC and a bifunctional chelator
  • Used in radiolabeling research
  • Supports research for tumor-assisted diagnosis
  • Purity of 99.50%
  • White to off-white solid appearance
  • Molecular weight of 814.88
  • Soluble in DMSO at concentrations of ≥ 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.