Learn More
Medchemexpress LLC HBED-CC-tris(tert-butyl ester) TFA | C40H57F3N2O12 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY153551A5MG
Encompass_Preferred
HBED-CC-tris(tert-butyl ester) TFA is a derivative of HBED-CC and a bifunctional chelator. It can be used in the research of radiolabeling and compounds for tumor-assisted diagnosis.
  • Can be used in the research of radiolabeling and compounds for tumor-assisted diagnosis.
  • Can be used in the research on the auxiliary diagnosis of prostate cancer when conjugated with PSMA and labeled with gallium isotopes.
  • Appears as a white to off-white solid.
  • Purity of 99.50%.
  • Intended for research use only.
Catalog No. 50-003-72791