Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HBED dihydrochloride | 35369-53-0 | MFCD04113603 | ≥95.0% | 461.34 g·mol⁻¹ | C20H26Cl2N2O6 | 250 MG
SDP

Catalog No. 5000303537
Encompass_Preferred

HBED dihydrochloride (CAS 35369-53-0) is an orally active, hexadentate phenolic aminocarboxylate iron chelator used in biochemical and pharmacological research to model iron binding and to study iron-overload in preclinical systems. Supplied as a white to off-white solid dihydrochloride salt, it is characterized by formula C20H26Cl2N2O6 and molecular weight 461.34 g·mol⁻¹.

  • Orally active hexadentate iron chelator.
  • Useful for in vitro and in vivo iron chelation studies.
  • White to off-white solid form, easy to handle and weigh.
  • Molecular formula C20H26Cl2N2O6; molecular weight 461.34 g·mol⁻¹.
  • High purity suitable for research applications (product documentation shows ≥95.0% with a reported 91.66% data-sheet variant).

Catalog No. 50-003-03537 Supplier Medchemexpress LLC Supplier No. 250MGHYW035520
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HBED dihydrochloride (CAS 35369-53-0) is an orally active, hexadentate phenolic aminocarboxylate iron chelator used in biochemical and pharmacological research to model iron binding and to study iron-overload in preclinical systems. Supplied as a white to off-white solid dihydrochloride salt, it is characterized by formula C20H26Cl2N2O6 and molecular weight 461.34 g·mol⁻¹.

  • Orally active hexadentate iron chelator.
  • Useful for in vitro and in vivo iron chelation studies.
  • White to off-white solid form, easy to handle and weigh.
  • Molecular formula C20H26Cl2N2O6; molecular weight 461.34 g·mol⁻¹.
  • High purity suitable for research applications (product documentation shows ≥95.0% with a reported 91.66% data-sheet variant).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.