Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ HBEGF (Human) ELISA Kit

Catalog No. 89101022 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
96 Assays
Sample Type:
CCS, Plasma (EDTA, Heparin), Serum
CCS, Plasma, Serum, Urine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sample Type
89-101-022 1 Kit CCS, Plasma (EDTA, Heparin), Serum
89-028-459 96 Assays CCS, Plasma, Serum, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-101-022 Supplier Abnova™ Supplier No. KA3343
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

In vitro enzyme-linked immunosorbent assay for the quantitative measurement of human HBEGF

  • Sample Type: Cell Culture Supernatant, Plasma, Serum, Urine
  • Sample Volume: 100μL

Quantification

Specifications

Calibration Range 15.6 to 1000pg/mL
Cross Reactivity Human
Description HBEGF (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human HBEGF.
Detection Method Colorimetric
Quantity 1 Kit
Sample Type CCS, Plasma (EDTA, Heparin), Serum
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type HBEGF (Human) ELISA kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.