Learn More
ABclonal Technology HBG1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A14989500UL
Encompass_Preferred
The gamma globin genes (HBG1 and HBG2) are normally expressed in the fetal liver, spleen and bone marrow. Two gamma chains together with two alpha chains constitute fetal hemoglobin (HbF) which is normally replaced by adult hemoglobin (HbA) at birth. In some beta-thalassemias and related conditions, gamma chain production continues into adulthood. The two types of gamma chains differ at residue 136 where glycine is found in the G-gamma product (HBG2) and alanine is found in the A-gamma product (HBG1). The former is predominant at birth. The order of the genes in the beta-globin cluster is 5-epsilon -- gamma-G -- gamma-A -- delta -- beta--3.
Catalog No. 50-001-05501