Learn More
Bulbman Inc HBO 50 W AC L2 Osram 69214
SDP
Catalog No. NC9206364
Encompass_Preferred
Miniature Mercury Vapor Light Bulb, 34 to 39V, 50w, It has an overall length of 2.1 in. and rated at 100 life hours
Catalog No. NC9206364 Supplier Bulbman Inc Supplier No. 105182
Miniature Mercury Vapor Light Bulb, 34 to 39V, 50w, It has an overall length of 2.1 in. and rated at 100 life hours