Learn More
SCANTIBODIES LABORATORY INC HBR PLUS (3KC545) 1.0G
Catalog No. NC2094015
Encompass
The 3KC545 Heterophilic Blocking Reagent Plus from Scantibodies Laboratory, Inc. was developed as an alternative for HBR-1. This product is compounded with immunoglobulins with different characteristics. Therefore, in addition to its active blocking characteristics, the special formulation and production procedures enhance its efficacy in its passive blocking ability as well. It has a concentration of 10.0 +/- 1.0 mg/ml.
Catalog No. NC2094015 Supplier SCANTIBODIES LABORATORY INC Supplier No. 3KC5451G
Add to Cart
Add to Cart
Request a Quote
The 3KC545 Heterophilic Blocking Reagent Plus from Scantibodies Laboratory, Inc. was developed as an alternative for HBR-1. This product is compounded with immunoglobulins with different characteristics. Therefore, in addition to its active blocking characteristics, the special formulation and production procedures enhance its efficacy in its passive blocking ability as well. It has a concentration of 10.0 +/- 1.0 mg/ml.