Learn More
SCANTIBODIES LABORATORY INC HBR 1, PURIFIED (3KC534) 1.0G
Catalog No. NC2200037
Encompass
The 3KC534 Heterophilic Blocking Reagent 1, Purified from Scantibodies Laboratory, Inc. is composed of murine derived proteins in a buffered salt solution, with a protein concentration of 2.1 mg/mL. Its physical and chemical properties are as a clear to opaque liquid, with no detectable odor.
This item is not returnable. View return policy
Catalog No. NC2200037 Supplier SCANTIBODIES LABORATORY INC Supplier No. 3KC5341G
The 3KC534 Heterophilic Blocking Reagent 1, Purified from Scantibodies Laboratory, Inc. is composed of murine derived proteins in a buffered salt solution, with a protein concentration of 2.1 mg/mL. Its physical and chemical properties are as a clear to opaque liquid, with no detectable odor.